Skip to Content
Merck
All Photos(1)

Documents

PZ0326

Sigma-Aldrich

PF-03549184

≥98% (HPLC)

Synonym(s):

N-(3′,4′-dichloro[1,1′-biphenyl]-4-yl)-4-hydroxy-2H-1,2-benzothiazine-3-carboxamide 1,1-dioxide, PF-9184

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H14Cl2N2O4S
CAS Number:
Molecular Weight:
461.32
UNSPSC Code:
12352202
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

InChI

1S/C21H14Cl2N2O4S/c22-16-10-7-13(11-17(16)23)12-5-8-14(9-6-12)25-19(21(24)27)20(26)15-3-1-2-4-18(15)30(25,28)29/h1-11,26H,(H2,24,27)

InChI key

YGTUCAUHSJOJCQ-UHFFFAOYSA-N

Biochem/physiol Actions

PF-03549184 (PF-9184) is a potent inhibitor of microsomal prostaglandin E synthase-1 (mPGES-1) with an IC50 value of 16.5 nM in recombinant human mPGES-1. mPEGS-1 is the terminal enzyme that synthesizes prostaglandin E2 (PGE2) from PGH2 to PGE2 downstream of cyclooxygenase-2 (COX-2). Its expression is induced in response to pro-inflammatory mediators, similar to COX-2. Unlike the effects of COX inhibition, PF-9184 does not inhibit the production of other PGH2-derived products such as 6-keto-PGFα or PGF2α. PF-9184 inhibits PGE2 production in cell based assays with IC50 ranges from 0.5 to 5 μM. IC50 concentrations for rhCOX1 and rHCOX2 are 118 μM and 236 μM, respectively.
PGE2 is involved in inflammation and is one of the most abundant prostanoid.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hui-Hua Chang et al.
Future medicinal chemistry, 3(15), 1909-1934 (2011-10-26)
Microsomal prostaglandin E synthase-1 (mPGES-1) is the terminal synthase responsible for the synthesis of the pro-tumorigenic prostaglandin E(2) (PGE(2)). mPGES-1 is overexpressed in a wide variety of cancers. Since its discovery in 1997 by Bengt Samuelsson and collaborators, the enzyme
Gabriel Mbalaviele et al.
Biochemical pharmacology, 79(10), 1445-1454 (2010-01-14)
Inflammation-induced microsomal prostaglandin E synthase-1 (mPGES-1) is the terminal enzyme that synthesizes prostaglandin E(2) (PGE(2)) downstream of cyclooxygenase-2 (COX-2). The efficacy of nonsteroidal anti-inflammatory drugs and COX-2 inhibitors in the treatment of the signs and symptoms of osteoarthritis, rheumatoid arthritis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service