Skip to Content
Merck
All Photos(1)

Documents

PIR02

Sigma-Aldrich

Inhibitor Resistant Genotyping PCR ReadyMix, ROX

with ROX for ABI instruments

Synonym(s):

Inhibitor Resistant Genotyping PCR ReadyMix

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106300
NACRES:
NA.55

Quality Level

form

liquid

usage

sufficient for 1250 reactions
sufficient for 250 reactions
sufficient for 5000 reactions

feature

Difficult Templates/Specialty Enzymes PCR
dNTPs included
hotstart

storage condition

protect from light

technique(s)

qPCR: suitable

color

light blue

input

crude DNA

compatibility

ABI 5700
ABI 7000
ABI 7300
ABI 7700
ABI 7900 HT
ABI 7900 HT Fast
ABI 7900
ABI StepOnePlus

detection method

probe-based

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Inhibitor Resistant Genotyping PCR Readymix is a 2X concentrated ready-to-use PCR master mix used for amplification of DNA templates that overcomes inhibitors often present in crude samples extracted from environmental specimens, plant tissues, or animal tissues. It is a versatile and robust PCR reagent that provides highly specific and efficient amplification for genotyping sequence variants including allelic discrimination of single nucleotide polymorphisms (SNPs) using a variety of fluorogenic probe chemistries, including TaqMan® hydrolysis probes. Inhibitor Resistant Genotyping PCR Readymix contains all required reaction components, except primers, probes, and DNA template. The light blue color of the inert tracer dye simplifies reaction assembly in white, or clear, plates and helps to minimize pipetting or mixing errors. It does not interfere with reporter dye fluorescence or affect the stability of the product.

A key component of Inhibitor Resistant Genotyping PCR ReadyMix is an ultra-pure, highly processive thermostable DNA polymerase that is combined with high avidity monoclonal antibodies. This proprietary polymerase mix is highly resistant to PCR inhibitors and provides an extremely stringent automatic hot-start allowing reaction assembly, and temporary storage, at room temperature prior to PCR amplification. Inhibitor Resistant Genotyping PCR Readymix is compatible with both fast and conventional PCR cycling protocols.

Features and Benefits

  • Assay results in as little as 33 minutes
  • Highly efficient and sensitive real-time PCR results
  • Little/no optimization required

Components

2X reaction buffer containing optimized concentrations of MgCl2, dNTPs (dATP, dCTP, dGTP, dTTP), hot-start DNA polymerase, ROX reference dye, insert blue qPCR dye, and stabilizers.

250 reactions* = 2 X 1.25 mL tubes
1250 reactions* = 10 X 1.25 mL tubes
5000 reactions* = 1 X 50 mL tube
*number of reactions based on a 20uL volume

Legal Information

ROX is a trademark of Applera Corporation or its subsidiaries in the US and/or certain other countries
ReadyMix is a trademark of Sigma-Aldrich Co. LLC
TaqMan is a registered trademark of Roche Molecular Systems, Inc.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service