Skip to Content
Merck
All Photos(1)

Key Documents

P3116

Sigma-Aldrich

[pSer392]-p53

>90% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202

Product Name

[pSer392]-p53, >90% (HPLC), solid

Assay

>90% (HPLC)

form

solid

mol wt

943

composition

Peptide content, ~80%

solubility

H2O: soluble

storage temp.

−20°C

Biochem/physiol Actions

Positive control peptide for the [Ser392]-p53 rabbit polyclonal antibody.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Mora-Castilla et al.
Cell death and differentiation, 17(6), 1025-1033 (2010-01-16)
Exposure of mouse embryonic stem (mES) cells to high concentrations of chemical nitric oxide (NO) donors promotes differentiation, but the mechanisms involved in this process at the gene expression level are poorly defined. In this study we report that culture

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service