Skip to Content
Merck
All Photos(1)

Key Documents

P0183

Finnpipette® pipette tips

Finntip 10, non-sterile

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121600
sterility:
non-sterile
feature:
barrier: no
packaging:
pack of 100 ea (Bulk packed)
manufacturer/tradename:
Finn 9402151

material

red
red tip

sterility

non-sterile

feature

barrier: no

packaging

pack of 100 ea (Bulk packed)

manufacturer/tradename

Finn 9402151

L

15 cm

maximum volume

10 mL

volume range

1-10 mL

Looking for similar products? Visit Product Comparison Guide

Other Notes

Browse all pipette tips

Legal Information

Finnpipette is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service