Skip to Content
Merck
All Photos(1)

Key Documents

N5663

Sigma-Aldrich

Monoclonal Anti-3-Nitrotyrosine antibody produced in mouse

~1 mg/mL, clone 2A12, purified immunoglobulin, buffered aqueous glycerol solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2A12, monoclonal

form

buffered aqueous glycerol solution

mol wt

antigen ~55 kDa

species reactivity

wide range

concentration

~1 mg/mL

technique(s)

western blot: 1:1,000 using mouse and rat brain

isotype

IgG1

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... NOS1(4842)

Looking for similar products? Visit Product Comparison Guide

Immunogen

3-Nitrotyrosine-KLH.

Physical form

Solution in 20 mM sodium phosphate, 150 mM sodium chloride, 50% glycerol, and 3 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service