Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
5000 ea | Please contact Customer Service for Availability | ₪2,311.00 |
About This Item
NACRES:
NB.15
UNSPSC Code:
41121706
Capacity:
3 mL
Material:
neck (rimless), polystyrene , round
Sterility:
non-sterile
Packaging:
case of × 5000 ea (10 bags of 500ea)
₪2,311.00
Please contact Customer Service for Availability
material
neck (rimless), polystyrene , round
sterility
non-sterile
packaging
case of × 5000 ea (10 bags of 500ea)
manufacturer/tradename
MTC™ Bio T1111
capacity
3 mL
diam. × L
12 mm × 60 mm
suitability
suitable for (cell culture)
General description
Bulk packed, economical test tubes for high volume users. Supplied non-sterile and without caps, available in either translucent polypropylene or crystal clear polystyrene. Caps can be ordered separately.
Test Tube / Culture Tube, 12x60mm, 3mL, PS, rimless, no cap, non-sterile, 10 bags of 500 tubes, 5000/cs
Features and Benefits
- Round Bottoms
- Non-Sterile
- Plug Caps Available Separately
- Resealable Zip Bags
- Non-Graduated
Legal Information
MTC is a trademark of MTC Bio
Still not finding the right product?
Explore all of our products under MTC™ Bio Disposable Test Tubes
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.