Skip to Content
Merck
All Photos(1)

Documents

MT30671554

Sigma-Aldrich

Mettler Toledo SevenDirect SD20 pH / ORP Meter Kit

electrode, InLab® Expert Pro-ISM

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115603
NACRES:
NB.61

AC/DC input

, universal plug set
220 V / 240 V

feature

electrode InLab® Expert Pro-ISM

packaging

pack of 1 kit

manufacturer/tradename

Mettler-Toledo 30671554

mV measuring range

-2,000.000-2,000.000 mV, accuracy: ±0.1, resolution: 1; 0.1

pH measuring range

-2.000-20.000, accuracy: ±0.002, resolution: 0.1
resolution: 0.001
resolution: 0.01

temperature measuring range

-30-130 °C, resolution: 0.1 °C

General description

Mastered in no Time
Large color touchscreen provides direct measurement and easy data entry. Instructions guide users through the sensor calibration process.
Tracks your Records
The instrument stores data like timestamp, sample and sensor ID with every measurement and prints or exports records automatically.
Tough Enough
Closed connector compartment and replaceable cover keep the instrument safe from dust and spills, even with sensors attached (IP54 in-use).
All-rounder pH Sensor for Liquid Samples
InLab Expert-Pro ISM is a robust and reliable pH/temperature sensor. Its polymer electrolyte prevents junction clogging and doesn′t require refill.
Electrode Arm for Efficient Operation
The EasyPlace electrode arm makes placing the sensor in the sample or buffer container easy. Dedicated positions for cleaning and storage support correct sensor handling. Vertical and horizontal movements facilitate precise positioning, even with one hand.
Protect Critical Settings
A two-level user management protects the most important settings. Operators retain the flexibility to individually set preferences like language and screen brightness.
Display Adapts to Your Needs
View detailed measurement information on one measurement screen - or focus on the result only and maximize readability with the uFocus mode.

Application

pH measurement

Features and Benefits

  • Large color touchscreen provides direct measurement and easy data entry.
  • The instrument stores data like timestamp, sample and sensor ID with every measurement and prints or exports records automatically.
  • Smart design and cover keeps the instrument safe from dust and spills
  • Low maintenance and robust pH/temperature sensor doesn′t require refill.
  • Electrode arm for efficient operation
  • Protect critical settings Protected with two-level user management
  • Display adapts to your needs to maximize readability

Components

Includes: Meter;Sensor; an efficient electrode arm; power supply; pH buffer sachets for initial calibration.

Legal Information

InLab is a registered trademark of Mettler-Toledo, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service