Skip to Content
Merck
All Photos(1)

Key Documents

M9065

Sigma-Aldrich

Methyl 14(S),15(S)-epoxy-13(S)-hydroxy-(5Z,8Z,11Z)-eicosatrienoate

≥95% (HPLC), hexane solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H34O4
Molecular Weight:
350.49
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥95% (HPLC)

form

hexane solution

functional group

ester

lipid type

fatty acid modified lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

CCCCCC1OC1[C@@H](O)\C=C/C\C=C/C\C=C/CCCC(=O)OC

InChI

1S/C21H34O4/c1-3-4-12-16-19-21(25-19)18(22)15-13-10-8-6-5-7-9-11-14-17-20(23)24-2/h6-9,13,15,18-19,21-22H,3-5,10-12,14,16-17H2,1-2H3/b8-6-,9-7-,15-13-/t18-,19-,21+/m0/s1

InChI key

DBXPKRNKMVEUMA-WBDQARBPSA-N

Other Notes

15-Lipoxygenase product of arachidonic acid, formed by a similar biosynthesis as hepoxilins

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Inhalation - STOT SE 3

Target Organs

Central nervous system, Nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-14.8 °F - closed cup

Flash Point(C)

-26 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service