Skip to Content
Merck
All Photos(1)

Key Documents

M3504

Sigma-Aldrich

L-Methioninamide hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H12N2OS · HCl
CAS Number:
Molecular Weight:
184.69
MDL number:
UNSPSC Code:
12352200

SMILES string

Cl.CSCC[C@H](N)C(N)=O


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W L Starnes et al.
European journal of biochemistry, 124(2), 363-370 (1982-05-17)
Human liver alanine aminopeptidase (EC 3.4.11.14; L-alpha-aminoacyl-peptide hydrolase) catalyzes the stepwise hydrolysis of methionyl-lysyl-bradykinin to yield methionine, lysine, and the limit nonapeptide, bradykinin which is resistant to further hydrolytic cleavage by this enzyme. Alanine aminopeptidase also catalyzes the hydrolysis of
M M Jones et al.
Anticancer research, 11(1), 449-453 (1991-01-01)
Six compounds containing a thioether group were examined as agents for the reduction of the nephrotoxicity caused by cisplatin (CDDP) in the rat. Of these, five were able to reduce the CDDP- induced nephrotoxicity when administered simultaneously with CDDP (8.0
András Láng et al.
Proteins, 58(3), 571-588 (2004-12-24)
The conformational space of the most biologically significant backbone folds of a suitable methionine peptide model was explored by density functional computational method. Using a medium [6-31G(d)] and a larger basis set [6-311++G(2d,2p)], the systematic exploration of low-energy backbone structures

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service