All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C10H15N3O4
CAS Number:
Molecular Weight:
241.24
EC Number:
MDL number:
UNSPSC Code:
12352204
Recommended Products
mp
217-219 °C (lit.)
storage temp.
−20°C
SMILES string
CC1=CN([C@H]2C[C@H](O)[C@@H](CO)O2)C(=O)N=C1N
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service