Skip to Content
Merck
All Photos(1)

Key Documents

M2257

Sigma-Aldrich

2′-O-Monosuccinyladenosine 3′:5′-cyclic monophosphate tyrosyl methyl ester

~95%

Synonym(s):

ScAMP-TME

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H27N6O11P
CAS Number:
Molecular Weight:
606.48
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:

Assay

~95%

form

solid

storage temp.

−20°C

SMILES string

COC(=O)C(N)Cc1ccc(OC(=O)CCC(=O)OC2C3OP(O)(=O)OCC3OC2n4cnc5c(N)ncnc45)cc1

InChI

1S/C24H27N6O11P/c1-36-24(33)14(25)8-12-2-4-13(5-3-12)38-16(31)6-7-17(32)40-20-19-15(9-37-42(34,35)41-19)39-23(20)30-11-29-18-21(26)27-10-28-22(18)30/h2-5,10-11,14-15,19-20,23H,6-9,25H2,1H3,(H,34,35)(H2,26,27,28)

InChI key

VCGXASLRSBTMRC-UHFFFAOYSA-N

Application

2′-O-Monosuccinyladenosine 3′:5′-cyclic monophosphate tyrosyl methyl ester (ScAMP-TME) is a high specific activity cAMP derivative that binds cAMP-binding antibodies. It may be used in assays, especially radioimmunoassys, for cAMP determination.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Schmidt et al.
Analytical biochemistry, 141(2), 499-502 (1984-09-01)
Tyrosine methyl esters of 2'-O-succinyl cyclic AMP and 2'-O-succinyl cyclic GMP were radioiodinated, and the products were purified by anion-exchange chromatography on polyethyleneimine-cellulose thin layers. Using 1.0 M LiCl for development, two major, immunologically active fractions (presumably the mono and
W F Oehlenschlager et al.
Journal of immunoassay, 11(1), 109-118 (1990-01-01)
2'-O-succinyladenosine 3':5'-cyclic monophosphate tyrosyl methyl ester (ScAMP-TME) and 2'-O-succinylguanosine 3':5'-cyclic monophosphate tyrosyl methyl ester (ScGMP-TME) were radioiodinated using chloramine T and Na125I. The resulting radiolabeled cyclic nucleotide derivatives, ScAMP-125I-TME and ScGMP-125I-TME, were subsequently purified by reverse-phase chromatography on Sep-Pak C18
T S Jap et al.
Zhonghua yi xue za zhi = Chinese medical journal; Free China ed, 50(2), 89-95 (1992-08-01)
Succinyl cyclic AMP, coupled with human albumin, was injected into rabbit to elicit antibodies to the cyclic nucleotide hapten. The succinyl cyclic nucleotides were conjugated to human albumin through a carbodiimide coupling procedure. The radioligand was prepared essentially by the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service