LUC60500-1
Phage Display ElectroCombo Pack, Trial size (3 strains × 4 rxns each)
sufficient for 12 reactions
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106615
Recommended Products
usage
sufficient for 12 reactions
availability
available only in (India)
shipped in
dry ice
General description
Phage Display ElectroCombo Pack (four transformations of each strain [TG1, SS320, ER2738]). MC1061 F- Electrocompetent Cells. A non-amber suppressor strain, identical to SS320 except that it lacks the F′episome. Highly efficient (≥ 4 × 1010 cfu/μ g) electrocompetent cells for general cloning or Phage Display. Note that as an F- strain, it cannot be used for re-infection by filamentous phage. This strain not included in the sample kit.TG1 Electrocompetent Cells. An amber suppressor strain (supE) prepared as highly efficient (≥ 4 × 1010 cfu/μg) electrocompetent cells for phage display library screening. These cells are used for phage display and protein expression.SS320 Electrocompetent Cells. A non-amber suppressor strain (sometimes called MC1061F′) prepared as highly efficient (≥ 4 × 1010 cfu/μg) electrocompetent cells for phage display library screening. These cells have the highest available transformation efficiency of any strain used for phage display.ER2738 Electrocompetent Cells. An amber suppressor strain (glnV) prepared as highly efficient (≥ 2 × 1010 cfu/μ g) electrocompetent cells for phage display library screening. This strain is recommended for use with New England Biolab′s Ph.D.™ Phage Display Kits.
Legal Information
Ph.D. is a trademark of New England Biolabs
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service