Skip to Content
Merck
All Photos(10)

Key Documents

HPA003316

Sigma-Aldrich

Anti-ELF3 antibody produced in rabbit

enhanced validation

Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-E74-like factor 3 antibody produced in rabbit, Anti-ets domain transcription factor, epithelial-specific antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
recombinant expression
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

QASPYHPGSCGAGAPSPGSSDVSTAGTGASRSSHSSDSGGSDVDLDPTDGKLFPSDGFRDCKKGDPKHGKRKRGRPRKLSKEYWDCLEGKKSKHAPRGTHLWEFIRDILIHP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ELF3(1999)

Looking for similar products? Visit Product Comparison Guide

General description

ELF3 (E74-like factor 3) gene is mapped to human chromosome 1q32.2. It is expressed in normal epithelia during keratinocyte differentiation. It is also found to be expressed in cell lines of epithelial origin and in organs with specialized epithelial cells such as lung, stomach, intestine and kidney.

Immunogen

E74-like factor 3 recombinant protein epitope signature tag (PrEST)

Application

Anti-ELF3 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

ELF3 (E74-like factor 3) gene encodes a protein belonging to the ETS (E26 transformation-specific) family of transcription factors. The protein may be involved in oral carcinogenesis and may serve as a potential biomarker of oral epithelial dysplasia and oral squamous cell carcinoma. ELF3 expression is found to be elevated in lung carcinoma and adenocarcinoma suggesting that the gene may be involved in lung tumorigenesis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST84790

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dandan Li et al.
Journal of experimental & clinical cancer research : CR, 41(1), 332-332 (2022-12-03)
The biological function of lncRNA ELF3-AS1 remains largely unknown in cancers. The cause of SNAI2 overexpression in tumor metastasis remains largely unclear. The molecular mechanisms underlying the high co-expression of antisense lncRNAs and adjacent protein-coding genes remains unclear. RNA-seq, CHIP
Ahmad A AbdulMajeed et al.
Annals of diagnostic pathology, 17(4), 331-340 (2013-05-07)
Early diagnosis and targeted therapy are crucial to mitigating the morbidity and mortality of oral squamous cell carcinoma. Among the potentially malignant oral disorders, epithelial dysplasia has known association with malignant transformation, but defensible gradation of dysplasia severity presents unmet
M J Tymms et al.
Oncogene, 15(20), 2449-2462 (1997-12-12)
The ETS family of genes are implicated in cancers such as Ewings sarcoma, acute myeloid leukemia and chronic myelomonocytic leukemia. Further, they have important functions in embryonic development. Hence, identification and characterization of members of this family are important. We

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service