Skip to Content
Merck

Skip To

GS74

GST M4-4, Recombinant Human

Synonym(s):

GTM4, glutathione S-transferase mu 4

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg
Please contact Customer Service for Availability
₪2,063.00

About This Item

NACRES:
NA.55
UNSPSC Code:
12352200
Form:
frozen liquid
Assay:
>95% (SDS-PAGE)
Biological source:
human
Recombinant:
expressed in E. coli
Mol wt:
26 kDa

₪2,063.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human

Quality Segment

recombinant

expressed in E. coli

assay

>95% (SDS-PAGE)

form

frozen liquid

specific activity

1.67 units/mg protein

mol wt

26 kDa

concentration

1.8 mg/mL

storage temp.

−70°C

Gene Information

human ... GSTM4(2948)

General description

using spectrophotometric determination of 1-chloro-2,4-dinitrobenzene (CDNB) conjugation with reduced glutathione (1 mM) in 100 mM NaPO4 (pH 6.5) at room temperature.

Biochem/physiol Actions

Cytosolic and membrane-bound forms of glutathione S-transferase are encoded by two distinct supergene families. At present, eight distinct classes of the soluble cytoplasmic mammalian glutathione S-transferases have been identified: alpha, kappa, mu, omega, pi, sigma, theta and zeta. This gene encodes a glutathione S-transferase that belongs to the mu class. The mu class of enzymes functions in the detoxification of electrophilic compounds, including carcinogens, therapeutic drugs, environmental toxins and products of oxidative stress, by conjugation with glutathione. The genes encoding the mu class of enzymes are organized in a gene cluster on chromosome 1p13.3 and are known to be highly polymorphic. These genetic variations can change an individual′s susceptibility to carcinogens and toxins as well as affect the toxicity and efficacy of certain drugs. Diversification of these genes has occurred in regions encoding substrate-binding domains, as well as in tissue expression patterns, to accommodate an increasing number of foreign compounds. Multiple transcript variants, each encoding a distinct protein isoform, have been identified.
Glutathione S-transferase mu 4 (GSTM4) is an enzyme that in humans is encoded by the GSTM4 gene. Glutathione S-transferases (GSTs) are a family of enzymes that play an important role in detoxification by catalyzing the conjugation of many hydrophobic and electrophilic compounds with reduced glutathione. Based on their biochemical, immunologic, and structural properties, cytosolic and membrane-bound forms of glutathione S-transferase are encoded by two distinct supergene families. At present, eight distinct classes of the soluble cytoplasmic mammalian glutathione S-transferases have been identified: alpha, kappa, mu, omega, pi, sigma, theta and zeta. The GSTs are thought to function in xenobiotic metabolism and play a role in susceptibility to cancer, and other diseases.

Preparation Note

The enzyme should be used by the end-user customer within 1 year of receipt.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
GS64GS65GS75
recombinant

expressed in E. coli

recombinant

expressed in E. coli

recombinant

expressed in E. coli

recombinant

expressed in E. coli

assay

>95% (SDS-PAGE)

assay

>95% (SDS-PAGE)

assay

>95% (SDS-PAGE)

assay

>95% (SDS-PAGE)

biological source

human

biological source

human

biological source

human

biological source

human

concentration

1.8 mg/mL

concentration

1.01 mg/mL

concentration

3.1 mg/mL

concentration

1.98 mg/mL

form

frozen liquid

form

frozen liquid

form

frozen liquid

form

frozen liquid

mol wt

26 kDa

mol wt

25 kDa

mol wt

26 kDa

mol wt

26 kDa


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters