Skip to Content
Merck
All Photos(1)

Key Documents

G4027

Sigma-Aldrich

D-(+)-Glyceric acid hemicalcium salt monohydrate

lyophilized powder

Synonym(s):

(R)-2,3-Dihydroxypropanoic acid hemicalcium salt, D-2,3-Dihydroxypropanoic acid hemicalcium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H5O4 · 0.5Ca · H2O
CAS Number:
Molecular Weight:
143.12
Beilstein:
6119062
MDL number:
UNSPSC Code:
12352200

form

lyophilized powder

mp

~141 °C (dec.)

SMILES string

[H]O[H].OC[C@@H](O)C(=O)O[Ca]OC(=O)[C@H](O)CO

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service