E5530
pTAC-MAT-Tag®-1 Expression Vector
bacterial vector for cytoplasmic expression of N-terminal MAT tag fusion proteins
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352200
Recommended Products
form
buffered aqueous solution
shipped in
dry ice
storage temp.
−20°C
Application
Cytoplasmic expression of N-terminal MAT-Tag® fusion proteins under control of the tac promoter.
Other Notes
Supplied with a pTAC-MAT-2+BAP Control Vector.
Browse additional application references in our FLAG® Literature portal.
Legal Information
FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany
MAT-Tag is a registered trademark of Merck KGaA, Darmstadt, Germany
related product
Storage Class Code
10 - Combustible liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Bacterial Expression Vectors: tac Promoter System
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service