Skip to Content
Merck
All Photos(1)

Key Documents

E147

Supelco

Ecgonine methyl ester-(N-methyl-d3) hydrochloride

Synonym(s):

(−)-3β-Hydroxy-1αH-5αH-tropane-2β-carboxylic acid methyl ester, 3-Hydroxy-8-(methyl-::FId::FN3)-8-azabicyclo[3.2.1]octane-2-carboxylic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14D3NO3 · HCl
CAS Number:
Molecular Weight:
238.73
UNSPSC Code:
41116107

grade

analytical standard

drug control

USDEA Schedule II; regulated under CDSA - not available from Sigma-Aldrich Canada

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

storage temp.

−20°C

InChI

1S/C10H17NO3.ClH/c1-11-6-3-4-7(11)9(8(12)5-6)10(13)14-2;/h6-9,12H,3-5H2,1-2H3;1H/t6-,7?,8?,9+;/m0./s1

InChI key

RLLMNWXOZDXKCQ-JVVAZNKBSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service