D5282
Lambda DNA EcoR I Digest
for DNA electrophoresis
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
UNSPSC Code:
12352200
NACRES:
NA.25
Recommended Products
form
liquid
usage
100 uses
suitability
suitable for electrophoresis (DNA)
storage temp.
−20°C
General description
Sigma′s Lambda DNA EcoR I Digest contains 6 fragments from 3,530 – 21,226 bp. Ten ul of the ladder should be diluted in gel loading buffer and then loaded in a single lane on an agarose or polyacrylamide gel.
Components
Sigma′s Lambda DNA EcoR I Digest is provided in a solution of 10 mM Tris-HCl (pH 8.0), with 1.0 mM EDTA.
Other Notes
Contains 6 fragments, 3,530–21,226 bp
For optimal resolution, the recommended agarose gel concentration is 0.7%.
Preparation Note
Bacteriophage lambda DNA was digested with EcoR I to completion, then purified by phenol extraction.
related product
Product No.
Description
Pricing
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Choose the appropriate markers and ladders for nucleic acid size determination of samples separated by electrophoresis. Determine size of DNA, RNA and PCR-generated fragments using agarose or polyacrylamide gels.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service