D2281
5-([4,6-Dichlorotriazin-2-yl]amino)fluorescein Hydrochloride–Celite®
suitable for protein labeling
Synonym(s):
DTAF
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
MDL number:
UNSPSC Code:
12171500
extent of labeling
~10% DTAF
suitability
suitable for protein labeling
storage temp.
2-8°C
SMILES string
Cl.OC(=O)c1cc(Nc2nc(Cl)nc(Cl)n2)ccc1C3=C4C=CC(=O)C=C4Oc5cc(O)ccc35
Legal Information
Celite is a registered trademark of Imerys Minerals California, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service