Skip to Content
Merck
All Photos(1)

Documents

CLS354526

Corning® BioCoat Fibronectin Cell Culture Flask

capacity 175 cm2, rectangular flask, Straight neck, cap, with Plug Seal Cap

Synonym(s):

cell culture flask, plastic cell culture flask, coated flask

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122104
NACRES:
NB.13

material

rectangular flask

feature

cap with Plug Seal Cap
Straight neck

packaging

pkg of 5 ea (1 pack of 5)

manufacturer/tradename

Corning 354526

capacity

175 cm2

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

Application

Cell culture

Legal Information

BioCoat is a trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service