CLS16864050
Corning® weighing bottle
diam. × H 40 mm × 50 mm
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
borosilicate glass bottle
clear bottle
cylindrical bottle
glass cap
wide-mouth bottle
feature
cap
packaging
case of 12 ea
pack of 6 ea
manufacturer/tradename
Corning 1686-4050
bottle neck diam.
~46 mm
capacity
45 mL
diam. × H
40 mm × 50 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
Legal Information
Corning is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service