Skip to Content
Merck
All Photos(1)

Key Documents

Psi-CRE

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

biological source

mouse embryo

packaging

tube of 5 μg 94090902-DNA-5UG
pkg of vial of cells 94090902-1VL

growth mode

Adherent

karyotype

Not specified

morphology

Fibroblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse NIH 3T3 transformed with Moloney murine leukaemia mutants

Cell Line Description

From a set of packaging cell lines useful for generating helper-free recombinant retroviruses with ectotropic host ranges. Two mutant Moloney murine leukaemia virus-derived proviral genomes carrying complementary mutations in the gag-pol or env regions were sequentially introduced into NIH 3T3 cells by co-transformation. Each genome contained a deletion of the Psi sequence necessary for the efficient encapsidation of retroviral genomes into virus particles and additional alterations at the 3′end of the provirus. Psi-CRE can be used to isolate clones that stably produce high titres (1000,000 cfu/ml) of recombinant retroviruses with ectotropic host ranges, viral producers derived from Psi-CRE do not transfer the packaging function or yield helper virus.

Application

Ecotropic helper-free recombinant retroviral packaging cell line. Valuable for `in vivo′ gene transfer studies aimed at cell lineage analysis and development of human gene replacement therapy.

Culture Medium

DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS) (NB. The results obtained by the depositor were gained using 10% Calf Serum (CS)).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4 x 10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5-10% CO2, 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service