Skip to Content
Merck
All Photos(1)

Documents

THM 7.3.5

Reactivity: Anti-NP (4-hydroxy-3 nitrophenacetyl), 87120201

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

biological source

Mouse + Human Transfectoma

ImmunoDon

Not applicable

Myeloma

J558L

Reactivity

Anti-NP (4-hydroxy-3 nitrophenacetyl)

technique(s)

cell culture | mammalian: suitable

isotype

IgM

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse J558L + Mouse VH gene + Human C(μ) gene

Cell Line Description

This human chimeric antibody is specifically directed against the hapten-4-hydroxy-3-nitrophenacetyl. V lambda of the endogenous light chain of plasmacytoma line complements VNP of transfected heavy chain to yield specific antibody. Gene marker: gpt.

Culture Medium

DMEM + 2mM glutamine + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Cultures from HPA Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service