Skip to Content
Merck
All Photos(1)

Documents

BB

87101201, fish trunk, Fibroblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

BB, 87101201

biological source

fish trunk

growth mode

Adherent

karyotype

2n = 60

morphology

Fibroblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Related Categories

Cell Line Origin

Fish Brown Bullhead posterior trunk tissue

Cell Line Description

Derived from the trunk tissue (excluding fins) of a 2 year old Brown Bullhead fish. These cells will tolerate a temperature range of 4-34°C. The optimal temperature range for cell proliferation is 25-30°C. BB is susceptible to infectious pancreatic necrosis virus and channel catfish virus.

Application

Virus studies: Infectious pancreatic necrotis virus and Channel catfish virus and channel catfish virus

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 using 0.25% trypsin/EDTA; 5% CO2; 23°C. Note: The cells do not grow well on glass. Fish cell lines detach easily during transit if the culture is too young. After resuscitation split cells 1:3 and culture for a

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service