Skip to Content
Merck
All Photos(1)

Documents

H129-1

14040203, mouse embryo, Small cells growing in large dense round colonies

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

H129-1, Haploid, embryonic stem cells, mouse, 14040203

biological source

mouse embryo

growth mode

Adherent

karyotype

Mixed (haploid and diploid)

morphology

Small cells growing in large dense round colonies

receptors

not known

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Description

Haploid embryonic mouse stem cell line established from inner cell mass derived from activated unfertilized oocytes from superovulated 129/Sv female mice.

Other Notes

Cultures from PHE Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.

Disclaimer

This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service