Skip to Content

CATA

Chloramphenicol Acetyltransferase (CAT) Reporter Gene Activity Detection Kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352200
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


usage

 kit sufficient for 100 tests

shipped in

dry ice

storage temp.

−20°C

Application

One of the most widely used reporter genes in mammalian expression systems is chloramphenicol acetyltransferase (CAT) from Escherichia coli. Mammalian cells have minimal CAT activity, resulting in high signal-to-background ratios for this reporter gene assay. Conventional CAT assays use a radioactive substrate and measure acetylated chloramphenicol. This carries all the disadvantages of radioactive assays plus another that is peculiar to the CAT reaction. Chloramphenicol has two free hydroxyl groups, but only the 3-OH is acetylated by CAT. The 3-acetate can rearrange to the 1-position non-enzymatically, and to the extent that it does, the 3-OH is open for further acetylation. The result is that three products are produced: two monoacetates and a diacetate. In addition, because the transacetylation is slow, the quantity of products is not a true measure of the CAT activity.

Features and Benefits

  • Provides a non-radioactive method that does not suffer from the disadvantages of the conventional assay
  • The substrate, a fluorescent derivative of 1-deoxychloramphenicol, has only a 3-hydroxyl group; only one enzymatic product is possible
  • Substrate and product are separated cleanly on TLC; UV illumination of the developed plate offers a quick qualitative indication of CAT activity
  • Product and substrate can be extracted from the TLC plate for a quantitative fluorimetric assay.

Legal Information

The BODIPY-labeled substrate and product are covered by U.S. Patent Nos. 5,262,545 and 5,364,764.


Kit Components Only

Product No.
Description

  • Detailed technical bulletin 1 ea

Kit Components Also Available Separately

Product No.
Description
SDS & Pricing

  • Acetyl-CoA (cofactor) 1 mL/vial
    SDS

  • BODIPY FL–1-deoxychloramphenicol 3-acetate (positive standard) 1 mL/vial
    SDS

  • BODIPY FL–1-deoxychloramphenicol (substrate) 1 mL/vial
    SDS

  • CAT enzyme (positive control) 250 U
    SDS

  • 5× lysis buffer 25 mL
    SDS



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library