Skip to Content

C3285

Complement C6 from human serum

80% (SDS-PAGE), >300,000 C6H50 units/mg protein

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352202
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

80% (SDS-PAGE)

specific activity

>300,000 C6H50 units/mg protein

concentration

1 mg/mL in PBS, pH 7.2

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... C6(729)

General description

Complement C6 is one of five complement cascade proteins that are incorporated into the lytic terminal complement complex. It is over 33% homologous with C7, including all 56 cysteine residues; C6 and C7 share the arrangement of LDL-receptor class A and B modules, the thrombospondin type I module at the C-terminus, and the two short consensus repeat modules.

The mature protein consists of 913 amino acid residues preceded by a 21-residue signal sequence.

Application

Complement C6 is one of the end terminals of the complement system contained in the membrane attack complex (MAC). It has been shown that complement C6 and the MAC are crucial for the development of arthritis in osteoarthritis and gouty arthritis. This could serve as a therapeutic target area for suppression of joint inflammation.

Analysis Note

Functionally active by a sensitive hemolytic assay.

Other Notes

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.


Storage Class

12 - Non Combustible Liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library