C1009
Z-Hydroxy-L-proline methyl ester
viscous liquid
Synonym(s):
N-Cbz-L-4-Hydroxyproline methyl ester, Z-HYP-OMe
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C14H17NO5
CAS Number:
Molecular Weight:
279.29
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
form
viscous liquid
storage temp.
−20°C
SMILES string
COC(=O)[C@@H]1C[C@@H](O)CN1C(=O)OCc2ccccc2
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service