BACSMLS
Bile Acid/Carnitine/Sterol Metabolite Library of Standards
Supplied by IROA Technologies
Synonym(s):
Mass Spectrometry Metabolite Library
About This Item
Recommended Products
form
solid
packaging
pkg of 1 plates
manufacturer/tradename
IROA Technologies
plate size
96 wells
shipped in
dry ice
storage temp.
−20°C
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
Features and Benefits
To view a complete list of compounds in the library for recent lots, download the plate maps below:
The complete plate map with Name, parent CID, molecular formula, molecular weight, CAS, ChEBI, HMDB ID, PubChem Compound and Substance ID are provided with the software download.
Preparation Note
Other Notes
Legal Information
related product
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service