Skip to Content
Merck

B170

BW373U86 hydrochloride

powder

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C27H37N3O2 · HCl
CAS Number:
Molecular Weight:
472.06
UNSPSC Code:
41116107
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

form

powder

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

color

tan

solubility

H2O: 4 mg/mL

format

neat

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Potent, nonpeptide δ-opioid receptor agonist.

Legal Information

Sold with the permission of Delta Pharmaceuticals Inc

Disclaimer

Hygroscopic, store desiccated

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ai Hashimoto et al.
Toxicologic pathology, 48(3), 509-523 (2020-01-04)
Macrophages and myofibroblasts are important in fibrogenesis. The cellular characteristics in pancreatic fibrosis remain to be investigated. Pancreatic fibrosis was induced in F344 rats by a single intravenous injection of dibutyltin dichloride. Histopathologically, the induced pancreatic fibrosis was divided into
K J Chang et al.
The Journal of pharmacology and experimental therapeutics, 267(2), 852-857 (1993-11-01)
Four different opioid receptor binding assays and three different isolated tissue studies were used to screen for delta receptor-selective nonpeptidic compounds. (+/-)-4-((alpha-R*)-alpha-((2S*,5R*)-4-Allyl-2,5- dimethyl-1-piperazinyl)-3-hydroxybenzyl)-N,N-diethylbenzamide (BW373U86) was a potent delta receptor-selective ligand in receptor binding assays. The Ki values were 1.8 +/-
S D Comer et al.
The Journal of pharmacology and experimental therapeutics, 267(2), 866-874 (1993-11-01)
Several opioid agonists were evaluated in pigeons trained to discriminate i.m. injections of sterile water from either the mu agonist morphine (5.6 mg/kg), the kappa agonist bremazocine (0.032mg/kg) or (+/-)-4-((alpha-R*)-alpha-((2S*,5R*)-4-allyl-2,5-dimethyl-1- piperazinyl)-3-hydroxybenzyl)-N,N-diethylbenzamide (BW373U86; 0.56 mg/kg). Pigeons were trained to peak one
S R Childers et al.
Molecular pharmacology, 44(4), 827-834 (1993-10-01)
BW373U86 is a potent and highly selective nonpeptidic agonist for delta-opioid receptors. To determine its ability to couple with G protein-linked second messenger systems, this study examined the effects of BW373U86 on the inhibition of adenylyl cyclase and the stimulation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service