All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C23H44N2O4
CAS Number:
Molecular Weight:
412.61
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
storage temp.
2-8°C
SMILES string
C1CCC(CC1)NC2CCCCC2.CCCC[C@H](NC(=O)OC(C)(C)C)C(O)=O
InChI
1S/C12H23N.C11H21NO4/c1-3-7-11(8-4-1)13-12-9-5-2-6-10-12;1-5-6-7-8(9(13)14)12-10(15)16-11(2,3)4/h11-13H,1-10H2;8H,5-7H2,1-4H3,(H,12,15)(H,13,14)/t;8-/m.0/s1
InChI key
BFEVJIUEBDZIJQ-WDBKTSHHSA-N
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service