Skip to Content
Merck

Skip To

A9481

AD 198

≥98% (HPLC), solid

Synonym(s):

N-Benzyladriamycin-14-valerate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C39H43NO12
CAS Number:
Molecular Weight:
717.76
UNSPSC Code:
12352200
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

form

solid

color

red

solubility

DMSO: >10 mg/mL, H2O: insoluble

storage temp.

2-8°C

SMILES string

CCCCC(=O)OCC(=O)[C@@]1(O)C[C@H](O[C@H]2C[C@H](NCc3ccccc3)[C@H](O)[C@H](C)O2)c4c(O)c5C(=O)c6c(OC)cccc6C(=O)c5c(O)c4C1

Biochem/physiol Actions

AD 198 is a PKC-delta activator.
AD 198 is a PKC-delta activator. Phospholipid scramblase 3 (PLS3) is an enzyme that plays a critical role in mitochondrial morphology, functions, and apoptotic response. During apoptosis, activated protein kinase C-delta (PKC-delta) translocates to mitochondria and phosphorylates PLS3. Used to show that PLS3 is a critical downstream effector of PKC-delta in AD 198-induced apoptosis.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



K Kashfi et al.
Biochemical pharmacology, 40(7), 1441-1448 (1990-10-01)
Adriamycin (ADR; doxorubicin) and its highly lipophilic, less toxic analogue N-benzyl-adriamycin-14-valerate (AD 198) were found to inhibit rat heart and liver carnitine palmitoyltransferases of both mitochondrial outer and inner membranes. The outer membrane enzyme was more sensitive to inhibition by
Yongwen He et al.
Cancer research, 65(21), 10016-10023 (2005-11-04)
Phospholipid scramblase 3 (PLS3) is an enzyme that plays a critical role in mitochondrial morphology, functions, and apoptotic response. During apoptosis, activated protein kinase C-delta (PKC-delta) translocates to mitochondria and phosphorylates PLS3. Here, we utilize an extranuclear-targeted anthracycline N-benzyladriamycin-14-valerate (AD198)
Leonard Lothstein et al.
Leukemia research, 31(8), 1085-1095 (2006-12-26)
Bcr-Abl activity in chronic myelogenous leukemia (CML) results in dysregulated cell proliferation and resistance against multiple cytotoxic agents due to the constitutive activation of proliferative signaling pathways. Currently, the most effective treatment of CML is the inhibition of Bcr-Abl activity