Skip to Content
Merck
All Photos(1)

Documents

A8627

Sigma-Aldrich

DL-Alanine methyl ester hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H9NO2 · HCl
CAS Number:
Molecular Weight:
139.58
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

storage temp.

−20°C

SMILES string

Cl.COC(=O)C(C)N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katarzyna Guzow et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 61(6), 1133-1140 (2005-03-03)
The photophysical properties of 3-[2-(4-diphenylaminophenyl)benzoxazol-5-yl]alanine methyl ester (1b) and its Boc derivative (1a) were studied in a series of solvents. Its UV-Vis absorption spectra are less sensitive to the solvent polarity than the corresponding fluorescence spectra which show pronounced solvatochromic
B J Bogitsh et al.
The Journal of parasitology, 77(2), 187-193 (1991-04-01)
Cultured Schistosoma mansoni schistosomula of various ages were exposed to several lysosomotropic agents. Weak bases such as chloroquine, ammonium chloride, and acridine orange caused gut swelling upon protonation. The latter compound fluoresced a bright orange indicating the acidic nature of
Erik Schwartz et al.
Chemical communications (Cambridge, England), (31)(31), 4675-4677 (2009-07-31)
Femtosecond vibrational pump-probe spectroscopy on beta-helical polyisocyanopeptides reveals vibrational self-trapping in the well-defined hydrogen-bonded side groups that is absent when non-hydrogen bonded monomers are mixed in.
Peter G W Gettins et al.
The Biochemical journal, 443(1), 65-73 (2011-12-21)
The LRP (low-density lipoprotein receptor-related protein) can bind a wide range of structurally diverse ligands to regions composed of clusters of ~40 residue Ca2+-dependent, disulfide-rich, CRs (complement-like repeats). Whereas lysine residues from the ligands have been implicated in binding, there
Jeanette Kheir et al.
The journal of physical chemistry. B, 117(10), 2872-2877 (2013-02-15)
In this study, the reactions of presolvated electrons with glycine methyl ester and N-acetylalanylalanine methyl ester (N-aAAMe) are investigated by electron spin resonance (ESR) spectroscopy and DFT calculations. Electrons were produced by γ-irradiation in neutral 7.5 M LiCl-D2O aqueous glasses

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service