Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
0.2 mL | Please contact Customer Service for Availability | ₪1,227.00 |
About This Item
biological source
mouse
Quality Segment
conjugate
unconjugated
antibody form
ascites fluid
antibody product type
primary antibodies
clone
AGP-47, monoclonal
contains
15 mM sodium azide
species reactivity
human, baboon (weakly)
should not react with
canine, bovine
technique(s)
dot blot: suitable, indirect ELISA: suitable, western blot: 1:10,000
isotype
IgG1
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... ORM1(5004)
General description
Immunogen
Biochem/physiol Actions
Physical form
Preparation Note
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| antibody form ascites fluid | antibody form ascites fluid | antibody form ascites fluid | antibody form ascites fluid |
| species reactivity human, baboon (weakly) | species reactivity rhesus monkey, baboon, human | species reactivity gibbon, baboon, human, rhesus monkey | species reactivity human |
| conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate unconjugated |
| biological source mouse | biological source mouse | biological source mouse | biological source mouse |
| Gene Information human ... ORM1(5004) | Gene Information human ... ALB(213) | Gene Information human ... ALB(213) | Gene Information human ... CRP(1401) |
| clone AGP-47, monoclonal | clone HSA-9, monoclonal | clone HSA-11, monoclonal | clone CRP-8, monoclonal |
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
10 - Combustible liquids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



