Skip to Content
Merck
All Photos(1)

Key Documents

A0456

Sigma-Aldrich

(2S,3R,4E)-2-Azido-4-octadecene-1,3-diol

Synonym(s):

D-Sphingosine azide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H35N3O2
CAS Number:
Molecular Weight:
325.49
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCC=C[C@@H](O)[C@H](CO)N=[N+]=[N-]

InChI

1S/C18H35N3O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-18(23)17(16-22)20-21-19/h14-15,17-18,22-23H,2-13,16H2,1H3/b15-14+/t17-,18+/m0/s1

InChI key

XAPVDQFHDYWVTA-KRWOKUGFSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G van Echten et al.
The Journal of biological chemistry, 265(16), 9333-9339 (1990-06-05)
Sphingolipid biosynthesis was studied in cultured murine cerebellar cells in the absence and presence of exogenous sphingosine homologues with different alkyl chain lengths (12, 18, and 24 carbon atoms). Labeling of cells with [14C]serine for 24 h indicated that endogenous

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service