Skip to Content
Merck
All Photos(1)

Key Documents

74304

Sigma-Aldrich

1,2-Dioleoyl-sn-glycero-3-phosphoric acid monosodium salt

≥98% (TLC)

Synonym(s):

3-sn-Phosphatidic acid, 1,2-dioleoyl monosodium salt, L-β,γ-Dioleoyl-α-phosphatidic acid monosodium salt, DOPA monosodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H72NaO8P
CAS Number:
Molecular Weight:
722.95
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

biological source

synthetic

Assay

≥98% (TLC)
≥98% (as oleic acid, GC)

form

solid

impurities

1.6-4.7% sodium

solubility

chloroform: 20 mg/mL, clear, colorless

functional group

phospholipid

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(O)=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O.[Na+]

InChI

1S/C39H73O8P.Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-38(40)45-35-37(36-46-48(42,43)44)47-39(41)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2;/h17-20,37H,3-16,21-36H2,1-2H3,(H2,42,43,44);/q;+1/p-1/b19-17-,20-18-;/t37-;/m1./s1

InChI key

JUKKLZHXFHPJSF-ZBFGHDQJSA-M

Biochem/physiol Actions

Dioleoylphosphatidic acid inhibits the spontaneous curvature of synthetic lipid monolayers in a dose-dependent manner.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shu Jie Li et al.
Biophysical chemistry, 109(1), 149-155 (2004-04-03)
We have investigated the effects of anionic dioleoylphosphatidic acid (DOPA) on the structure and phase behavior of dipalmitoleoylphosphatidylethanolamine (DPOPE) membranes by small-angle X-ray scattering. The results of X-ray diffraction experiments indicate that an L(alpha) to H(II) phase transition in DPOPE

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service