Skip to Content
Merck

Skip To

73645

Trifluoroacetic acid

≥99.5%

Synonym(s):

TFA

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
CF3COOH
CAS Number:
Molecular Weight:
114.02
EC Number:
200-929-3
UNSPSC Code:
12190000
PubChem Substance ID:
Beilstein/REAXYS Number:
742035
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor density

3.9 (vs air)

vapor pressure

97.5 mmHg ( 20 °C)

assay

≥99.5% (T), ≥99.5%

impurities

≤0.05% water

refractive index

n20/D 1.3 (lit.)

bp

72.4 °C (lit.)

mp

−15.4 °C (lit.)

density

1.489 g/mL at 20 °C (lit.)

suitability

in accordance for sequence analysis test

SMILES string

OC(C(F)(F)F)=O

InChI

1S/C2HF3O2/c3-2(4,5)1(6)7/h(H,6,7)

InChI key

DTQVDTLACAAQTR-UHFFFAOYSA-N



Still not finding the right product?

Explore all of our products under Trifluoroacetic acid


pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1A

Storage Class

8A - Combustible corrosive hazardous materials

wgk

WGK 2

flash_point_f

>212.0 °F - Pensky-Martens closed cup

flash_point_c

> 100 °C - Pensky-Martens closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Russell J Chedgy et al.
Phytochemistry, 113, 149-159 (2015-01-07)
Salicinoids are phenolic glycosides (PGs) characteristic of the Salicaceae and are known defenses against insect herbivory. Common examples are salicin, salicortin, tremuloidin, and tremulacin, which accumulate to high concentrations in the leaves and bark of willows and poplars. Although their