Skip to Content
Merck

71776

N-(+)-Biotinyl-3-aminopropylammonium trifluoroacetate

≥97.0% (HPLC)

Synonym(s):

N-(3-Aminopropyl)biotinamide trifluoroacetate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C13H24N4O2S · C2HF3O2
CAS Number:
Molecular Weight:
414.44
UNSPSC Code:
12352202
NACRES:
NA.25
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
8665808
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥97.0% (HPLC)

form

powder

storage temp.

2-8°C

SMILES string

OC(=O)C(F)(F)F.[H][C@]12CS[C@@H](CCCCC(=O)NCCCN)[C@@]1([H])NC(=O)N2

InChI

1S/C13H24N4O2S.C2HF3O2/c14-6-3-7-15-11(18)5-2-1-4-10-12-9(8-20-10)16-13(19)17-12;3-2(4,5)1(6)7/h9-10,12H,1-8,14H2,(H,15,18)(H2,16,17,19);(H,6,7)/t9-,10-,12-;/m0./s1

InChI key

XIFKYHZIURRTHB-SVMJMOIZSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Novel reagent for labeling of electrophilic derivatives with biotin


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library