Skip to Content
Merck

52335

N-Hexadecylpyrene-1-sulfonamide

suitable for fluorescence, ≥90% (TLC)

Synonym(s):

N-(Pyrene-1-sulfonyl)hexadecylamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C32H43NO2S
Molecular Weight:
505.75
UNSPSC Code:
12352200
PubChem Substance ID:
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥90% (TLC)

solubility

DMSO: soluble, chloroform: soluble, methanol: soluble

fluorescence

λex 350 nm; λem 400 nm in methanol

suitability

suitable for fluorescence

SMILES string

CCCCCCCCCCCCCCCCNS(=O)(=O)c1ccc2ccc3cccc4ccc1c2c34

InChI

1S/C32H43NO2S/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-25-33-36(34,35)30-24-22-28-20-19-26-17-16-18-27-21-23-29(30)32(28)31(26)27/h16-24,33H,2-15,25H2,1H3

InChI key

FAXRWJMTLJBKAD-UHFFFAOYSA-N

Application

N-Hexadecylpyrene-1-sulfonamide may be used in the analysis of drug delivery devices for lipophilic pyrenyl compounds.


Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library