Skip to Content
Merck
All Photos(1)

Key Documents

49845

Sigma-Aldrich

ASB-C80

≥95% (TLC)

Synonym(s):

3-{N,N-Dimethyl-N-[3-(4-octylbenzoylamino)propyl]ammonio}propanesulfonate, N,N-Dimethyl-N-[3-[(4-octylbenzoyl)amino]propyl]-3-sulfo-1-propanaminium inner salt, C80 Detergent

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H40N2O4S
CAS Number:
Molecular Weight:
440.64
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:

description

zwitterionic

Assay

≥95% (TLC)

mol wt

440.64 g/mol

SMILES string

CCCCCCCCc1ccc(cc1)C(=O)NCCC[N+](C)(C)CCCS([O-])(=O)=O

InChI

1S/C23H40N2O4S/c1-4-5-6-7-8-9-12-21-13-15-22(16-14-21)23(26)24-17-10-18-25(2,3)19-11-20-30(27,28)29/h13-16H,4-12,17-20H2,1-3H3,(H-,24,26,27,28,29)

InChI key

AQVJFUZGGMXMED-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Macri et al.
Electrophoresis, 21(9), 1685-1693 (2000-06-28)
We evaluated the use of the alkyaryl amidosulfobetaine zwitterionic detergent, designated as C8psi, to facilitate the solubilization of cardiac subcellular, membrane-associated proteins. Hearts from 7-week-old male Sprague-Dawley rats were isolated, and the left ventricles dissected and subsequently homogenized. The sarcolemma
Robert Henningsen et al.
Proteomics, 2(11), 1479-1488 (2002-11-21)
Comparative analysis has long been utilized in biological research to interpret protein interactions in both drug naïve versus drug challenged and normal versus diseased tissues. The technology of proteomics today allows researchers to provide insight into old and still open

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service