Skip to Content
Merck
All Photos(1)

Key Documents

49701

Sigma-Aldrich

7-(Dimethylamino)-1-(2-propynyloxy)-phenoxazin-3-one

suitable for fluorescence

Synonym(s):

7-(Dimethylamino)-1-(2-propyn-1-yloxy)-3H-phenoxazin-3-one, Red Oxazin-X-Alkyne

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H14N2O3
CAS Number:
Molecular Weight:
294.30
Beilstein:
19893167
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

solid

Quality Level

solubility

methanol: 1 mg/mL, clear

fluorescence

λex 570 nm; λem 625 nm±10 nm in methanol

suitability

suitable for fluorescence

storage temp.

−20°C

SMILES string

O=C1C=C2OC3=CC(N(C)C)=CC=C3N=C2C(OCC#C)=C1

InChI

1S/C17H14N2O3/c1-4-7-21-15-9-12(20)10-16-17(15)18-13-6-5-11(19(2)3)8-14(13)22-16/h1,5-6,8-10H,7H2,2-3H3

InChI key

ZHJHFGKPSPYQGZ-UHFFFAOYSA-N

General description

a clickable dye

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service