Skip to Content
Merck

Skip To

41882

Atto 430LS maleimide

suitable for fluorescence

Sign In to View Organizational & Contract Pricing.

About This Item

NACRES:
NA.32
UNSPSC Code:
12352200


mol wt

Mw 711 g/mol

manufacturer/tradename

ATTO-TEC GmbH

fluorescence

λex 433 nm; λem 540.0 nm±10 nm in PBS, pH 7.4

suitability

suitable for fluorescence

storage temp.

−20°C

General description

Atto 430LS is a new fluorescent label featuring an extraordinary large Stokes-Shift of 114 nm. Thus its emission spectrum is almost completely separated from its absorption spectrum, making the dye highly suitable for multiplexing experiments. Atto 430LS is very hydrophilic and shows excellent water solubility. The dye exhibits a high fluorescence quantum yield, which is only slightly reduced after conjugation to biomolecules, e.g. proteins, even at high degrees of labeling (DOL).
Atto 430LS is an anionic dye. After conjugation to a substrate the dye carries a net electrical charge of -1. The fluorescence is excited most efficiently in the range 400 - 460 nm.

find more information here

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.


Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters