Skip to Content
Merck
All Photos(1)

Key Documents

09969

Sigma-Aldrich

SQ-NHS

suitable for fluorescence, >50% (coupling to amines)

Synonym(s):

Squarain-carboxylate N-succinimidyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H32ClN3O6
CAS Number:
Molecular Weight:
614.09
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.32

Quality Level

Assay

>50% (coupling to amines)

solubility

DMF: soluble

fluorescence

λex 600 nm; λem 645 nm in 0.1 M phosphate pH 7.0

suitability

suitable for fluorescence

storage temp.

2-8°C

InChI

1S/C34H32ClN3O6/c1-7-37-25-10-8-18(32(43)44-38-28(39)12-13-29(38)40)14-22(25)34(4,5)27(37)17-21-30(41)20(31(21)42)16-26-33(2,3)23-15-19(35)9-11-24(23)36(26)6/h8-11,14-17H,7,12-13H2,1-6H3

InChI key

RRLJXAKQFSXTLC-UHFFFAOYSA-N

Application

SQ-NHS contains a reactive N-hydroxysuccinimide group, which allows for coupling with amino groups. When conjugated with proteins, SQ-NHS demonstrates a high quantum yield with an absorption maximum at 640 nm and a nanosecond lifetime. Additionally, in the physiological pH range (6-9), the absorbance, emission, and lifetime of SQ-NHS are pH-independent. It is water insoluble .

related product

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service