Skip to Content
Merck

08340

5-Aminolevulinic acid hydrochloride

≥97.0% (AT)

Synonym(s):

δ-Aminolevulinic acid hydrochloride, 5-Amino-4-oxopentanoic acid hydrochloride, 5-Aminolaevulinic acid hydrochloride, ALA

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Linear Formula:
NH2CH2C(O)CH2CH2COOH · HCl
CAS Number:
Molecular Weight:
167.59
EC Number:
226-679-5
UNSPSC Code:
12352200
PubChem Substance ID:
Beilstein/REAXYS Number:
3690651
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

assay

≥97.0% (AT)

form

powder or crystals

impurities

≤1% water

color

white to faint beige

mp

~150 °C (dec.)

anion traces

sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤50 mg/kg, Cd: ≤5 mg/kg, Co: ≤5 mg/kg, Cr: ≤5 mg/kg, Cu: ≤5 mg/kg, Fe: ≤10 mg/kg, K: ≤100 mg/kg, Mg: ≤5 mg/kg, Mn: ≤5 mg/kg, Na: ≤50 mg/kg, Ni: ≤60 mg/kg, Pb: ≤5 mg/kg, Zn: ≤20 mg/kg

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

Cl[H].NCC(=O)CCC(O)=O

InChI

1S/C5H9NO3.ClH/c6-3-4(7)1-2-5(8)9;/h1-3,6H2,(H,8,9);1H

InChI key

ZLHFONARZHCSET-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Intermediate in heme biosynthesis.

Other Notes

Substrate for the assay of γ-aminolevulinic acid dehydratase

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D. Appleton et al.
Journal of the Chemical Society. Perkin Transactions 1, 89-89 (1998)
Ruth A Elliott et al.
The Journal of urology, 186(6), 2455-2462 (2011-10-25)
Artificial sweeteners augment bladder contraction. We hypothesized that artificial sweeteners activate sweet taste receptors in the bladder. Thus, we investigated the expression of sweet taste receptors in human and rat bladders. Sections of human and rat bladders were cut from
A Petzold et al.
Journal of neuroimmunology, 138(1-2), 45-48 (2003-05-14)
Quantification of neurodegeneration in animal models is typically assessed by time-consuming and observer-dependent immunocytochemistry. This study aimed to investigate if newly developed ELISA techniques could provide an observer-independent, cost-effective and time-saving tool for this purpose. Neurofilament heavy chain (NfH(SM135)), astrocytic
Fábio Trindade et al.
PloS one, 15(9), e0237000-e0237000 (2020-09-19)
Currently, aortic valve replacement is the only treatment capable of relieving left ventricle pressure overload in patients with severe aortic stenosis. It aims to improve cardiac function and revert hypertrophy, by triggering myocardial reverse remodeling. Despite immediately relieving afterload, reverse
David J Berkoff et al.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society, 34(4), 616-622 (2015-09-30)
This study evaluated if inhibiting IL1-β activity with an IL1-receptor antagonist (IL1-RA) will prevent pathologic changes commonly seen in tendinopathy. Thirty-six Sprague-Dawley retired-breeder rats were divided into three groups having weekly bilateral patellar tendon injections: CON (0.1 ml Saline), CAR

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service