Skip to Content
Merck

Skip To

07876

Atto Rho3B Biotin

BioReagent, suitable for fluorescence

Sign In to View Organizational & Contract Pricing.

About This Item

NACRES:
NA.32
UNSPSC Code:
12352200


product line

BioReagent

manufacturer/tradename

ATTO-TEC GmbH

fluorescence

λex 564 nm; λem 592 nm

suitability

suitable for fluorescence

storage temp.

−20°C

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library






Global Trade Item Number

SKUGTIN
147214-25G04061838737267

Questions

Reviews

No rating value

Active Filters