Skip to Content
Merck
All Photos(1)

Key Documents

05717

Sigma-Aldrich

1,2-Dioctadecanoyl-sn-glycero-3-phospho-rac-(1-glycerol) ammonium salt

≥98.0% (TLC)

Synonym(s):

1,2-Dioctadecanoyl-sn-glycero-3-phospho-(1′-rac-glycerol) ammonium salt, 1,2-Distearoyl-sn-glycero-3-phosphoglycerol ammonium salt, L-α-Phosphatidyl-DL-glycerol, distearoyl ammonium salt, DSPG-NH4, PG(18:0/18:0)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H86NO10P
CAS Number:
Molecular Weight:
796.11
MDL number:
UNSPSC Code:
51191904
PubChem Substance ID:
NACRES:
NA.85

biological source

synthetic

Assay

≥98.0% (TLC)

form

powder

functional group

ester

lipid type

phosphoglycerides

storage temp.

−20°C

SMILES string

N.CCCCCCCCCCCCCCCCCC(=O)OC[C@H](COP(O)(=O)OCC(O)CO)OC(=O)CCCCCCCCCCCCCCCCC

InChI

1S/C42H83O10P.H3N/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-41(45)49-37-40(38-51-53(47,48)50-36-39(44)35-43)52-42(46)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2;/h39-40,43-44H,3-38H2,1-2H3,(H,47,48);1H3/t39?,40-;/m1./s1

InChI key

PJOUKPGQSVEHLZ-QTOMIGAPSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

DSPG is the most efficient phosphatidylglycerol in enhancing respiratory mucus clearance in cystic fibrosis.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria Bretner et al.
Antiviral chemistry & chemotherapy, 16(5), 315-326 (2005-10-26)
To improve anti-helical activity of analogues of 1H-benzotriazole and 1H-benzimidazole their N-alkyl derivatives were synthesized and tested for antihelicase activity against enzymes of selected Flaviviridae including hepatitis C virus (HCV), West Nile virus (WNV), Dengue virus (DENV) and Japanese encephalitis
Pin-Chien Huang et al.
PloS one, 9(4), e95090-e95090 (2014-04-30)
Developing retinas display retinal waves, the patterned spontaneous activity essential for circuit refinement. During the first postnatal week in rodents, retinal waves are mediated by synaptic transmission between starburst amacrine cells (SACs) and retinal ganglion cells (RGCs). The neuromodulator adenosine
Ying Zhu et al.
Current protocols in neuroscience, 91(1), e86-e86 (2020-01-17)
Combining immunological and molecular biological methods, the antibody-based proximity ligation assay (PLA) has been used for more than a decade to detect and quantify protein-protein interactions, protein modification, and protein expression in situ, including in brain tissue. However, the transfer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service