Skip to Content
Merck
All Photos(1)

Documents

334278

Sigma-Aldrich

Molecular sieves, 3 Å

pellets, 3.2 mm

Synonym(s):

Molecular Sieve UOP Type 3Å

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KnNa12-n[(AlO2)12(SiO2)12] · xH2O
CAS Number:
MDL number:
UNSPSC Code:
23201100
eCl@ss:
32110204
NACRES:
NB.24

form

pellets

Quality Level

particle size

3.2 mm

pore size

3 Å

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service