Skip to Content
Merck
All Photos(1)

Documents

16446

Sigma-Aldrich

Xylenes

Synonym(s):

Xylene mixture of isomers

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(CH3)2
CAS Number:
Molecular Weight:
106.17
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

vapor density

3.7 (vs air)

vapor pressure

18 mmHg ( 37.7 °C)

Assay

≥99.0%

form

liquid

expl. lim.

7 %

impurities

≤0.002% non-volatile matter
≥96% total isomers (GC)

refractive index

n20/D 1.4960-1.4990
n20/D 1.497 (lit.)

bp

137-140 °C (lit.)

mp

-34 °C

density

0.86 g/mL at 25 °C (lit.)

SMILES string

Cc1ccc(C)cc1.Cc2cccc(C)c2.Cc3ccccc3C

InChI

1S/3C8H10/c1-7-3-5-8(2)6-4-7;1-7-4-3-5-8(2)6-7;1-7-5-3-4-6-8(7)2/h3*3-6H,1-2H3

InChI key

MVZVDAGWAAZJPE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Pertains only in Germany: unversteuert

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

77.0 °F - closed cup

Flash Point(C)

25 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qiming Wang et al.
Nature communications, 5, 4899-4899 (2014-09-17)
Cephalopods can display dazzling patterns of colours by selectively contracting muscles to reversibly activate chromatophores--pigment-containing cells under their skins. Inspired by this novel colouring strategy found in nature, we design an electro-mechano-chemically responsive elastomer system that can exhibit a wide
Jovita M Saquing et al.
Environmental science & technology, 44(3), 1123-1129 (2009-12-31)
Desorption rates of toluene, o-xylene and tetrachloroethene from individual municipal solid waste components [high-density polyethylene (HDPE); poly(vinyl chloride) (PVC); office paper; newsprint; and rabbit food, a model food and yard waste] were determined. Effects of sorbent and sorbate properties, solvent
Lian He et al.
Journal of environmental sciences (China), 23(1), 160-165 (2011-04-12)
The activities of CeO2 nanocubes calcined at different temperatures were tested for catalytic oxidation of o-xylene. Using CeO2 nanocubes as catalysts, complete catalytic oxidation of o-xylene was achieved below 210 degrees C. The CeO2 nanomaterials were characterized by means of
Jennifer V Littlejohns et al.
Journal of hazardous materials, 175(1-3), 872-882 (2009-12-05)
A dynamic mathematical model has been developed to predict the performance of a stirred tank, solid-liquid two-phase partitioning bioreactor (SL-TPPB) for the treatment of benzene, toluene, ethylbenzene and o-xylene (BTEX) contaminated gases. The SL-TPPB system consists of an aqueous phase
Cheng-Xiong Yang et al.
Analytical chemistry, 83(18), 7144-7150 (2011-08-04)
The diverse structures and pore topologies, accessible cages and tunnels, and high surface areas make metal-organic frameworks attractive as novel media in separation sciences. Here we report the slurry-packed MIL-101(Cr) column for high-performance liquid chromatographic separation of substituted aromatics. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service