About This Item
Recommended Products
grade
reagent grade
vapor density
2.58 (vs air)
vapor pressure
15.6 mmHg ( 20 °C)
Assay
≥98.0%
form
liquid
autoignition temp.
778 °F
expl. lim.
3.4 %
refractive index
n20/D 1.391 (lit.)
bp
114-115 °C (lit.)
mp
−90 °C (lit.)
solubility
acetone: soluble(lit.)
alcohol: soluble(lit.)
water: slightly soluble(lit.)
density
1.045 g/mL at 25 °C (lit.)
SMILES string
CC[N+]([O-])=O
InChI
1S/C2H5NO2/c1-2-3(4)5/h2H2,1H3
InChI key
MCSAJNNLRCFZED-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
pK of nitroethane is 8.60.4
Nitroethane is an aprotic solvent with high polarity[2], which on oxidation with glucose oxidase gives nitrite, acetaldehyde and hydrogen peroxide as the major products.[3]
Application
Other Notes
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 1B - Flam. Liq. 3 - Muta. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
87.8 °F - closed cup
Flash Point(C)
31 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service