Key Documents
Z801321
Hellma® absorption cuvettes, standard cells, Macro
Suprasil® 300 quartz, spectral range 200-3500 nm, pathlength 1 mm, chamber volume 700 μL
About This Item
NACRES:
SB.32
material
Suprasil® 300 quartz
description
2 polished windows
manufacturer/tradename
Hellma 404.000-QX
parameter
200-3500 nm spectral range
H × W × D
47.5 mm × 23.6 mm × 7.5 m
chamber volume
700 μL
pathlength
1 mm
General description
with 2 PTFE stoppers
Packaging
Sold as packages containing a single cell (1EA = 1 cell)
Legal Information
Hellma is a registered trademark of Hellma GmbH
Suprasil is a registered trademark of HERAEUS QUARZGLAS GMBH & CO.
Choose from one of the most recent versions:
Certificates of Analysis (COA)